Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Transglutaminase II, Polyclonal

Catalog No. 616465500UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
61-646-5500UL 500 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-646-5500UL Supplier MilliporeSigma™ Supplier No. 616465500UL

Rabbit Polyclonal Antibody

Specifically detects Transglutaminase II Clone: in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

Transglutaminases are a family of enzymes that catalyze calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. Transglutaminase II has been implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transglutaminase II
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, 0.2% BSA, pH 7.4
Host Species Rabbit
Immunogen full-length, recombinant, human cellular/tissue transglutaminase II
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽77-85kDa transglutaminase II protein.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.