Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transient Receptor Potential Channel 7 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB15562200UL
Description
This Anti-Transient Receptor Potential Channel 7 Antibody is validated for use in immunocytochemistry, western blotting for the detection of Transient Receptor Potential Channel 7.
Specifications
Transient Receptor Potential Channel 7 | |
Polyclonal | |
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 200 uL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). | |
Rabbit | |
Immunoaffinity purified | |
Primary | |
Mouse, Rat | |
Purified |
Immunocytochemistry, Western Blot | |
Unconjugated | |
TRPM7; TRP-PLIK; LTRPC7; LTrpC7; FLJ20117; CHAK; FLJ25718; CHAK1; EC 2.7.11.1 | |
Purified peptide from the C-terminus of human TRPM7 (Accession number Q96QT4). | |
200 μL | |
NM_017672.3 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction