Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transient Receptor Potential Channel 7 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1556250UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1556250UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1556250UL Supplier MilliporeSigma Supplier No. AB1556250UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Transient Receptor Potential Channel 7 Antibody is an antibody against Transient Receptor Potential Channel 7 for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transient Receptor Potential Channel 7
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).
Gene Symbols TRPM7; TRP-PLIK; LTRPC7; LTrpC7; FLJ20117; CHAK; FLJ25718; CHAK1; EC 2.7.11.1
Host Species Rabbit
Immunogen Purified peptide from the C-terminus of human TRPM7 (Accession number Q96QT4).
Purification Method Immunoaffinity purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_017672.3
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.