Learn More
Transient Receptor Potential Vanilloid 4 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Transient Receptor Potential Vanilloid 4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 x |
| Gene Symbols | TRPV4; VROAC; TRP12; VRL2; VR-OAC; OTRPC4; VRL-2; TrpV4 |
| Host Species | Rabbit |
| Immunogen | Purified peptide from amino acids 853-871 of rat TRVP4 (Accession number Q9ERZ8). |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.