Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transthyretin Mouse anti-Human, Clone: 7G8-E11-E7, MilliporeSigma™

Catalog No. MAB10762MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10762MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10762MI Supplier MilliporeSigma Supplier No. MAB10762
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Transthyretin Antibody, clone 7G8-E11-E7 is validated for use in ELISA, western blotting for the detection of Transthyretin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transthyretin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7G8-E11-E7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_000362.1
Gene Symbols TTR, ATTR, PALB, Prealbumin, TBPA, transthyretin
Host Species Mouse
Immunogen Purified human Transthyretin.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.