Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-TRAP-1 Clone: TR-1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1276
Description
Specifically detects TRAP-1 Clone: TR-1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Heat shock protein 75kDa, mitochondrial (UniProt Q12931; also known as HSP 75, TNFR-associated protein 1, TRAP-1, Tumor necrosis factor type 1 receptor-associated protein) is encoded by the TRAP1 (also known as HSP75) gene (Gene ID 10131) in human. The mammalian HSP70 family consists of the mitochondrial HSP75 (TRAP-1), the endoplasmic HSP78, the cytoplasmic HSC70 (a.k.a. HSC73 and HSP73) and its inducible counterpart HSP72 (a.k.a. HSP70). TRAP-1 regulates a metabolic switch between oxidative phosphorylation and aerobic glycolysis. TRAP1-deficiency results in enhanced mitochondrial respiration and fatty acid oxidation, as well as increased cellular accumulation of tricarboxylic acid cycle intermediates, ATP, and reactive oxygen species species. TRAP-1-deficient cells display suppressed glucose metabolism and strikingly enhanced invasiveness, implicating a tumor suppressor role of TRAP-1.
Specifications
TRAP-1 | |
Monoclonal | |
Unconjugated | |
Q12931 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
TR-1 | |
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
TRAP1, HSP75 | |
Recombinant protein corresponding to human TRAP-1. | |
100 μg | |
Signaling | |
NP_057376.2 | |
2°C to 8°C for one year from date of shipment | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction