Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anti-Trastuzumab (Anti-Idiotype) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9547SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9547SP 25 μg
MAB9547100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9547SP Supplier R&D Systems Supplier No. MAB9547SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Trastuzumab Monoclonal specifically detects Trastuzumab in Multi-Species samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Trastuzumab
Applications Western Blot, ELISA
Classification Monoclonal
Clone 972618
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Host Species Mouse
Immunogen Full-length Trastuzumab
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Test Specificity Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.