Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAX Mouse, Unlabeled, Clone: 56, BD

Catalog No. BDB611358 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611358 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611358 Supplier BD Biosciences Supplier No. 611358

Mouse Monoclonal Antibody

RNA-binding proteins, such as Translin, have numerous functions that include mRNA transport and translational regulation, as well as DNA rearrangement and repair. Translin has a ring-shaped structure that allows binding to single stranded DNA ends. It is localized in the cytoplasm and nucleus, however it does not have a nuclear targeting motif. Translin-associated factor X (TRAX), a member of the Translin protein family, contains six potential PKC phosphorylation sites, eight CK2 phosphorylation sites, and one N-glycosylation site. TRAX can bind to Translin, contains a bipartite nuclear targeting motif, and has a heptad repeat of hydrophobic amino acids which may be a leucine zipper domain. This latter domain may be important for the formation of TRAX homodimers or heterodimers with Translin. Both TRAX and Translin are co-expressed in many tissues and form a complex with the GS1-DNA complex which is enriched in brain. Thus, TRAX may be involved with the nuclear transport of Translin, along with other important roles in regulating RNA transport and translation in a fashion similar to other RNA-binding proteins.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAX
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 56
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human TRAX aa. 1-207
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.