Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TRB3 (1-145), Polyclonal

Catalog No. ST1032100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1032100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1032100UL Supplier MilliporeSigma™ Supplier No. ST1032100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TRB3 (1-145) Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunoprecipitation

TRB3 is a protein that is reported to disrupt insulin signaling by binding directly to Akt and blocking activation of the kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRB3 (1-145)
Applications Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen a recombinant protein consisting of amino acids 1-145 of mouse TRB3, fused to GST
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽45kDa TRB3 protein in HepG2 cells.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.