Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRBC1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP150786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150786 0.1 mg
NB018671 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP150786 Supplier Novus Biologicals Supplier No. NBP150786

Rabbit Polyclonal Antibody

TRBC1 Polyclonal antibody specifically detects TRBC1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRBC1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 15 - 25 ug/mL, Immunoprecipitation 10 - 20 ug/mL
Formulation PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin 300
Gene Accession No. P01850
Gene Alias BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1
Gene Symbols TRBC1
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 114 of human TRBC1
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28639
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.