Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trefoil Factor 3 (TFF3) Rabbit anti-Canine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS185
Description
This Anti-Trefoil Factor 3 (TFF3) antibody is validated for use in western blotting, immunohistochemistry (Paraffin) and Multiplexing for the detection of Trefoil Factor 3 (TFF3).
Specifications
Trefoil Factor 3 (TFF3) | |
Polyclonal | |
Purified rabbit polyclonal serum in PBS containing 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Canine |
Multiplex, Immunoassay, Western Blot | |
Unconjugated | |
TFF3; ITF | |
Thio-tagged recombinant protein corresponding to the mature chain of canine Trefoil factor 3 (TFF3). | |
100 μg | |
NP_001002990 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction