Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREML1/TLT-1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP145208
Description
TREML1/TLT-1 Polyclonal specifically detects TREML1/TLT-1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
TREML1/TLT-1 | |
Polyclonal | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:2000 | |
NP_835468.1 | |
TREML1 | |
Peptide with sequence CGPAQNPPNNQTPS corresponding to C-Terminus according to NP_835468.1. | |
0.1 mg | |
Primary | |
This antibody may cross-react with mouse TREML1 (GeneID: 71326; MW: 34.28 kDa). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
dJ238O23.3, GLTL1825, PRO3438, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
340205 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction