Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREX1 Mouse anti-Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB611987 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611987 150 μg
BDB611986 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611987 Supplier BD Biosciences Supplier No. 611987
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA replication, repair, and recombination requires the excision of nucleotides from the DNA 3′ termini. Many 3′ to 5′ exonucleases have been identified which catalyze the excision of monophosphates from the 3′ termini of DNA. TREX1 and TREX2 are 3′ to 5′ exonucleases that contain three conserved exonuclease active site motifs (EASM) that may produce exonuclease activity. TREX1 and TREX2 are most closely related to the proofreading exonucleases of the bacterial replicative DNA polymerases and the RNase T enzymes. Recombinant expression of TREX1 and TREX2 demonstrates that they have exonuclease activity when oligonucleotide is present. TREX1 shows the greatest exonuclease activity with partial duplex DNA, and no activity with single-stranded RNA or an RNA-DNA partial duplex. In addition, reconstitution of TREX1 with DNA polymerase β and DNA ligase III-XRCC1 facilitates accurate rejoining of a 3′ mismatched base residue at a single-strand break. Thus, TREX1 and TREX2 are 3′ to 5′ exonucleases that may be important for excision of nucleotides during DNA replication, repair, and recombination.

Host Species: Mouse
Clone: 29
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Mouse TRE1 aa. 82-179

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TREX1
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse TRE1 aa. 82-179
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.