Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF1 Mouse anti-Human, Mouse, Rat, Hamster, Monkey, Clone: BED5 57-6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04638
Description
This Anti-TRF1 Antibody, clone BED5 57-6 is validated for use in western blot, immunofluorescence for the detection of TRF1.
Specifications
| TRF1 | |
| Monoclonal | |
| Unconjugated | |
| NP_059523 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Immunofluorescence, Western Blot | |
| BED5 57-6 | |
| Affinity purified mouse ascites in PBS with 0.05% NaN3. | |
| TERF1, telomeric repeat binding factor (NIMA-interacting) 1, FLJ41416, PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1 | |
| Human TRF1 protein | |
| 100 μg | |
| Hamster, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction