Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE210
Description
Use Anti-TRF1 Antibody (Rabbit Polyclonal Antibody) validated in western blotting to detect TRF1 also known as Telomeric repeat-binding factor 1.
Specifications
TRF1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Western Blot | |
Unconjugated | |
TRF1; Terf1 | |
GST-tagged recombinant protein corresponding to mouse TRF1. | |
100 μL | |
NP_033378 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction