Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF2 Mouse anti-Human, Rat, Clone: 4A794, MilliporeSigma™

Catalog No. 05521MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-521-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-521-MI Supplier MilliporeSigma Supplier No. 05521
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TRF2 Antibody, clone 4A794 is a Mouse Monoclonal Antibody for detection of TRF2 also known as Telomeric repeat-binding factor 1 and has been tested in immunocytochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4A794
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 150mM Tris/glycine, 0.05% sodium azide, 5mg/ml BSA before the addition of glycerol to 30%
Gene Accession No. NM_005652
Gene Symbols TERF2, TRF2, TRBF2
Host Species Mouse
Immunogen His-tagged, fusion-protein, corresponding to full-length TRF2 (Telomeric Repeat binding Factor 2), expressed in baculovirus
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.