Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tribbles homolog 3 (Drosophila), Mouse, Clone: 2G5, Abnova™

Catalog No. 89001329 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-329 Supplier Abnova Corporation Supplier No. H00057761M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRIB3.

The protein encoded by this gene is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1. [provided by RefSeq

Sequence: PDRATAVATASRLGPYVLLEPEEGGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMH

Specifications

Antigen tribbles homolog 3 (Drosophila)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRIB3.
Formulation PBS with no preservative; pH 7.4
Gene TRIB3
Gene Accession No. BC027484
Gene Alias C20orf97/NIPK/SINK/SKIP3/TRB3
Gene Symbols TRIB3
Host Species Mouse
Immunogen TRIB3 (AAH27484, 56 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57761
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.