Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIC-A Rabbit anti-Human, Rat, Mouse, Chimpanzee, Polyclonal, MilliporeSigma™

Catalog No. 061005MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-100-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-100-5MI Supplier MilliporeSigma Supplier No. 061005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TRIC-A Antibody detects level of TRIC-A and has been published and validated for use in western blot, immunohistochemistry and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIC-A
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in PBS containing 0.05% sodium azide.
Gene Symbols TRIC-A; MGC3169; TRICA
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human TRIC-A.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_076979.1
Target Species Chimpanzee, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.