Learn More
tripartite motif-containing 16, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TRIM16 protein.
Supplier: Abnova Corporation H00010626D01P
Description
This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined. [provided by RefSeq
Sequence: MAELDLMAPGPLPRATAQPPAPLSPDSGSPSPDSGSASPVEEEDVGSSEKLGRETEEQDSDSAEQGDPAGEGKEVLCDFCLDDTRRVKAVKSCLTCMVNYCEEHLQPHQVNIKLQSHLLTEPVKDHNWRYCPAHHSPLSAFCCPDQQCICQDCCQEHSGHTIVSLDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMAAISNTVQFLEEYCKFKNTEDITFPSVYVGLKDKLSGIRKVITESTVHLIQLLENYKKKLQEFSKEEEYDIRTQVSAVVQRKYWTSKPEPSTREQFLQYAYDITFDPDTAHKYLRLQEENRKVTNTTPWEHPYPDLPSRFLHWRQVLSQQSLYLHRYYFEVEIFGAGTYVGLTCKGIDRKGEERNSCISGNNFSWSLQWNGKEFTAWYSDMETPLKAGPFRRLGVYIDFPGGILSFYGVEYDTMTLVHKFACKFSEPVYAAFWLSKKENAIRIVDLGEEPEKPAPSLVGTAPSpecifications
tripartite motif-containing 16 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human TRIM16 protein. | |
TRIM16 | |
EBBP | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006470.3 | |
TRIM16 | |
TRIM16 (NP_006461.3, 1 a.a. ∼ 564 a.a) full-length human protein. | |
100 μg | |
Transcription Regulation | |
10626 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.