Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220709
Description
TRIM2 Polyclonal specifically detects TRIM2 in Human samples. It is validated for Western Blot.Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| KIAA0517tripartite motif protein 2, RING finger protein 86, RNF86tripartite motif protein TRIM2, tripartite motif containing 2, tripartite motif-containing 2, tripartite motif-containing protein 2 | |
| Rabbit | |
| 82 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 0.28mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9C040 | |
| TRIM2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TRIM2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 23321 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction