Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 21, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018283 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-283 Supplier Abnova Corporation Supplier No. H00006737D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRIM21 protein.

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined. [provided by RefSeq

Sequence: MASAARLTMMWEEVTCPICLDPFVEPVSIECGHSFCQECISQVGKGGGSVCPVCRQRFLLKNLRPNRQLANMVNNLKEISQEAREGTQGERCAVHGERLHLFCEKDGKALCWVCAQSRKHRDHAMVPLEEAAQEYQEKLQVALGELRRKQELAEKLEVEIAIKRADWKKTVETQKSRIHAEFVQQKNFLVEEEQRQLQELEKDEREQLRILGEKEAKLAQQSQALQELISELDRRCHSSALELLQEVIIVLERSESWNLKDLDITSPELRSVCHVPGLKKMLRTCAVHITLDPDTANPWLILSEDRRQVRLGDTQQSIPGNEERFDSYPMVLGAQHFHSGKHYWEVDVTGKEAWDLGVCRDSVRRKGHFLLSSKSGFWTIWLWNKQKYEAGTYPQTPLHLQVPPCQVGIFLDYEAGMVSFYNITDHGSLIYSFSECAFTGPLRPFFSPGFNDGGKNTAPLTLCPLNIGSQGSTDY

Specifications

Antigen tripartite motif-containing 21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRIM21 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRIM21
Gene Accession No. BC010861
Gene Alias RNF81/RO52/SSA/SSA1
Gene Symbols TRIM21
Host Species Rabbit
Immunogen TRIM21 (NP_003132.2, 1 a.a. ∼ 475 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6737
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.