Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 23, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018015 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-015 Supplier Abnova Corporation Supplier No. H00000373D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRIM23 protein.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein is also a member of the ADP ribosylation factor family of guanine nucleotide-binding family of proteins. Its carboxy terminus contains an ADP-ribosylation factor domain and a guanine nucleotide binding site, while the amino terminus contains a GTPase activating protein domain which acts on the guanine nucleotide binding site. The protein localizes to lysosomes and the Golgi apparatus. It plays a role in the formation of intracellular transport vesicles, their movement from one compartment to another, and phopholipase D activation. Three alternatively spliced transcript variants for this gene have been described. [provided by RefSeq

Sequence: MATLVVNKLGAGVDSGRQGSRGTAVVKVLECGVCEDVFSLQGDKVPRLLLCGHTVCHDCLTRLPLHGRAIRCPFDRQVTDLGDSGVWGLKKNFALLELLERLQNGPIGQYGAAEESIGISGESIIRCDEDEAHLASVYCTVCATHLCSECSQVTHSTKTLAKHRRVPLADKPHEKTMCSQHQVHAIEFVCLEEGCQTSPLMCCVCKEYGKHQGHKHSVLEPEANQIRASILDMAHCIRTFTEEISDYSRKLVGIVQHIEGGEQIVEDGIGMAHTEHVPGTAENARSCIRAYFYDLHETLCRQEEMALSVVDAHVREKLIWLRQQQEDMTILLSEVSAACLHCEKTLQQDDCRVVLAKQEITRLLETLQKQQQQFTEVADHIQLDASIPVTFTKDNRVHIGPKMEIRVVTLGLDGAGKTTILFKLKQDEFMQPIPTIGFNVETVEYKNLKFTIWDVGGKHKLRPLWKHYYLNTQAVVFVVDSSHRDRISEAHSELAKLLTEKELRDALLLIFANKQDVAGALSVEEITELLSLHKLCCGRSWYIQGCDARSGMGLYEGLDWLSRQLVAAGVLDVA

Specifications

Antigen tripartite motif-containing 23
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRIM23 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRIM23
Gene Accession No. NM_001656
Gene Alias ARD1/ARFD1/RNF46
Gene Symbols TRIM23
Host Species Rabbit
Immunogen TRIM23 (NP_001647.1, 1 a.a. ∼ 574 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 373
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.