Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 25, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-571 Supplier Abnova Corporation Supplier No. H00007706D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRIM25 protein.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. The presence of potential DNA-binding and dimerization-transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to estrogen, and it is thought to mediate estrogen actions in breast cancer as a primary response gene. [provided by RefSeq

Sequence: MAELCPLAEELSCSICLEPFKEPVTTPCGHNFCGSCLNETWAVQGSPYLCPQCRAVYQARPQLHKNTVLCNVVEQFLQADLAREPPADVWTPPARASAPSPNAQVACDHCLKEAAVKTCLVCMASFCQEHLQPHFDSPAFQDHPLQPPVRDLLRRKCSQHNRLREFFCPEHSECICHICLVEHKTCSPASLSQASADLEATLRHKLTVMYSQINGASRALDDVRNRQQDVRMTANRKVEQLQQEYTEMKALLDASETTSTRKIKEEEKRVNSKFDTIYQILLKKKSEIQTLKEEIEQSLTKRDEFEFLEKASKLRGISTKPVYIPEVELNHKLIKGIHQSTIDLKNELKQCIGRLQELTPSSGDPGEHDPASTHKSTRPVKKVSKEEKKSKKPPPVPALPSKLPTFGAPEQLVDLKQAGLEAAAKATSSHPNSTSLKAKVLETFLAKSRPELLEYYIKVILDYNTAHNKVALSECYTVASVAEMPQNYRPHPQRFTYCSQVLGLHCYKKGIHYWEVELQKNNFCGVGICYGSMNRQGPESRLGRNSASWCVEWFNTKISAWHNNVEKTLPSTKATRVGVLLNCDHGFVIFFAVADKVHLMYKFRVDFTEALYPAFWVFSAGATLSICSPK

Specifications

Antigen tripartite motif-containing 25
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRIM25 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRIM25
Gene Accession No. BC016924.1
Gene Alias EFP/RNF147/Z147/ZNF147
Gene Symbols TRIM25
Host Species Rabbit
Immunogen TRIM25 (AAH16924.1, 1 a.a. ∼ 630 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 7706
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.