Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti- TRIM3 Clone: 13D12.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC945
Description
Specifically detects TRIM3 Clone: 13D12.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
Tripartite motif-containing protein 3 (UniProt O75382; also known as Brain-expressed RING finger protein, RING finger protein 22, Ring finger protein 97, Tripartite motif protein TRIM3, Tripartite motif-containing 3) is encoded by the TRIM3 (also known as BERP, HAC1, RNF22, RNF97) gene (Gene ID 10612) in human. TRIM3 belongs to a family of more than 70 TRIM/RBCC proteins characterized by their shared tripartite motif structure, also known as the RBCC (RING finger, B-box, and coiled-coil) domain. The B-box and coiled coil regions are believed to participate in protein-protein interactions and formation of macromolecular complexes. TRIM proteins are involved in diverse cellular processes, including cell proliferation, differentiation, oncogenesis, and apoptosis. The presence of a RING domain suggests that TRIM proteins function as E3 ubiquitin ligases. TRIM3 is reported to catalyze the transfer of ubiquitin from UbcH5a to p21, a protein necessary for the proliferation of a subset of PDGF (platelet-derived growth factor)-transformed proneural glioma cells. Loss of TRIM3 heterozygosity is found in more than 10% of human GBMs (glioblastomas), while reducing TRIM3 expression facilitates tumor development and accelerates the progression of oligodendroglioma in a mouse model.
Specifications
TRIM3 | |
Monoclonal | |
Unconjugated | |
O75382 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Expected to react with all four spliced isoforms. | |
2°C to 8°C for one year from date of shipment | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
13D12.1 | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
TRIM3, BERP, RNF22, RNF97 | |
GST-tagged recombinant protein corresponding to the internal fragment of human TRIM3. | |
100 μg | |
Apoptosis, Cancer | |
NP_001234935 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction