Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 33, Mouse, Clone: 6F4, Abnova™

Catalog No. 89001284 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-284 Supplier Abnova Corporation Supplier No. H00051592M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRIM33.

The protein encoded by this gene is thought to be a transcriptional corepressor. However, molecules that interact with this protein have not yet been identified. The protein is a member of the tripartite motif family. This motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Three alternatively spliced transcript variants for this gene have been described, however, the full-length nature of one variant has not been determined. [provided by RefSeq

Sequence: VKKKLQKKHSQHYQIPDDFVADVRLIFKNCERFNEMMKVVQVYADTQEINLKADSEVAQAGKAVALYFEDKLTEIYSDRTFAPLPEFEQEEDDGEVTEDS

Specifications

Antigen tripartite motif-containing 33
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRIM33.
Formulation PBS with no preservative; pH 7.4
Gene TRIM33
Gene Accession No. NM_015906
Gene Alias FLJ32925/PTC7/RFG7/TF1G/TIF1G/TIF1GAMMA/TIFGAMMA
Gene Symbols TRIM33
Host Species Mouse
Immunogen TRIM33 (NP_056990, 1006 a.a. ∼ 1105 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.