Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 37, Mouse, Clone: 2D11, Abnova™

Catalog No. 89014152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-152 Supplier Abnova Corporation Supplier No. H00004591M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRIM37.

This gene encodes a member of the tripartite motif (TRIM) family, whose members are involved in diverse cellular functions such as developmental patterning and oncogenesis. The TRIM motif includes zinc-binding domains, a RING finger region, a B-box motif and a coiled-coil domain. The RING finger and B-box domains chelate zinc and might be involved in protein-protein and/or protein-nucleic acid interactions. The gene mutations are associated with mulibrey (muscle-liver-brain-eye) nanism, an autosomal recessive disorder that involves several tissues of mesodermal origin. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq

Sequence: GHLEGLQMTDLENNSETGELQPVLPEGASAAPEEGMSSDSDIECDTENEEQEEHTSVGGFHDSFMVMTQPPDEDTHSSFPDGEQIGPEDLSFNTDENSGR

Specifications

Antigen tripartite motif-containing 37
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRIM37.
Formulation PBS with no preservative; pH 7.4
Gene TRIM37
Gene Accession No. NM_015294
Gene Alias KIAA0898/MUL/POB1/TEF3
Gene Symbols TRIM37
Host Species Mouse
Immunogen TRIM37 (NP_056109, 865 a.a. ∼ 964 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4591
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.