Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM47 Mouse anti-Human, Clone: 3C8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA522843

Catalog No. PIMA522843


Add to cart

Description

Description

Peptide Sequence: PLPHPFSPTV GVCLEYADRA LAFYAVRDGK MSLLRRLKAS RPRRGGIPAS PIDPFQSRLD SHFAGLFTHR LKPAFFLESV DAHLQIGPLK KSCISVLKRR

Gene overexpressed in astrocytoma
Specifications

Specifications

TRIM47
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q96LD4
GOA, RNF100, RING finger protein 100, gene overexpressed in astrocytoma protein, tripartite motif-containing 47, tripartite motif-containing protein 47
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
91107
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3C8
Unconjugated
TRIM47
Liquid
TRIM47
TRIM47 (NP_258411, 539 a.a. ∽ 638 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit