Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM5 alpha Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABF206MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF206MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF206MI Supplier MilliporeSigma Supplier No. ABF206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Tripartite motif-containing protein 5 using this rabbit polyclonal antibody, Anti-TRIM5 alpha Antibody validated for use in western blotting, immunohistochemistry and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM5 alpha
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols TRIM5; RNF88
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the intermediate domain of human TRIM5 alpha.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.