Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tripartite motif-containing 55, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-780 Supplier Abnova Corporation Supplier No. H00084675D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRIM55 protein.

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: MSASLNYKSFSKEQQTMDNLEKQLICPICLEMFTKPVVILPCQHNLCRKCASDIFQASNPYLPTRGGTTMASGGRFRCPSCRHEVVLDRHGVYGLQRNLLVENIIDIYKQESTRPEKKSDQPMCEEHEEERINIYCLNCEVPTCSLCKVFGAHKDCQVAPLTHVFQRQKSELSDGIAILVGSNDRVQGVISQLEDTCKTIEECCRKQKQELCEKFDYLYGILEERKNEMTQVITRTQEEKLEHVRALIKKYSDHLENVSKLVESGIQFMDEPEMAVFLQNAKTLLKKISEASKAFQMEKIEHGYENMNHFTVNLNREEKIIREIDFYREDEDEEEEEGGEGEKEGEGEVGGEAVEVEEVENVQTEFPGEDENPEKASELSQVELQAAPGALPVSSPEPPPALPPAADAPVTQIGFEAPPLQGQAAAPASGSGADSEPARHIFSFSWLNSLNE

Specifications

Antigen tripartite motif-containing 55
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRIM55 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRIM55
Gene Accession No. NM_184086.1
Gene Alias MURF-2/RNF29
Gene Symbols TRIM55
Host Species Rabbit
Immunogen TRIM55 (NP_908974.1, 1 a.a. ∼ 452 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Primary or Secondary Primary
Gene ID (Entrez) 84675
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.