Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tripartite motif-containing 56, Mouse, Clone: 4C5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TRIM56.
Supplier: Abnova Corporation H00081844M01
Description
Sequence: NSVVICDGLGQVVGEYKGPGLHGCQPGSVSVDKKGYIFLTLREVNKVVILDPKGSLLGDFLTAYHGLEKPRVTTMVDGRYLVVSLSNGTIHIFRVRSPDSSpecifications
tripartite motif-containing 56 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_030961 | |
TRIM56 | |
TRIM56 (NP_112223, 656 a.a. ∼ 755 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Ubiquitin/Proteasome | |
81844 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
4C5 | |
Mouse monoclonal antibody raised against a partial recombinant TRIM56. | |
TRIM56 | |
DKFZp667O116/FLJ35608/RNF109 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction