Learn More
tripartite motif-containing 6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TRIM6.
Supplier: Abnova Corporation H00117854A01
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A read-through transcript transcribed from this gene and TRIM34 has been observed. [provided by RefSeq
Sequence: DVTERSEFWTLRKPEALPTKLRSMFRAPDLKRMLRVCRELTDVQSYWVDVTLNPHTANLNLVLAKNRRQVRFVGAKVSGPSCLEKHYDCSVLGSQHFSSGSpecifications
| tripartite motif-containing 6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TRIM6. | |
| TRIM6 | |
| RNF89 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human, Rat | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001003818 | |
| TRIM6 | |
| TRIM6 (NP_001003818, 282 a.a. ∼ 381 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 117854 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.