Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM6-TRIM34 readthrough transcript, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004836 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-836 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-836 Supplier Abnova Corporation Supplier No. H00445372A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TRIM6-TRIM34.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This gene represents a read-through transcript transcribed from genes TRIM6 and TRIM34, and it was described as a splice variant of TRIM34. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. [provided by RefSeq

Sequence: LQMFRELTAVRCYWVDVTLNSVNLNLNLVLSEDQRQVISVPIWPFQCYNYGVLGSQYFSSGKHYWEVDVSKKTAWILGVYCRTYSRHMKYVVRRCANRQN

Specifications

Antigen TRIM6-TRIM34 readthrough transcript
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TRIM6-TRIM34.
Formulation 50% glycerol
Gene TRIM6-TRIM34
Gene Accession No. NM_001003819
Gene Alias IFP1/RNF21/TRIM34
Gene Symbols TRIM6-TRIM34
Host Species Mouse
Immunogen TRIM6-TRIM34 (NP_001003819, 641 a.a. ∼ 740 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 445372
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.