Learn More
tripartite motif-containing 63 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle. [provided by RefSeq]
Specifications
Specifications
| Antigen | tripartite motif-containing 63 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant TRIM63. |
| Formulation | 50% glycerol |
| Gene | TRIM63 |
| Gene Accession No. | NM_032588 |
| Gene Alias | FLJ32380/IRF/MURF1/MURF2/RNF28/SMRZ |
| Gene Symbols | TRIM63 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.