Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM8, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017174 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-174 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-174 Supplier Abnova Corporation Supplier No. H00081603B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TRIM8 protein.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes. [provided by RefSeq

Sequence: MAENWKNCFEEELICPICLHVFVEPVQLPCKHNFCRGCIGEAWAKDSGLVRCPECNQAYNQKPGLEKNLKLTNIVEKFNALHVEKPPAALHCVFCRRGPPLPAQKVCLRCEAPCCQSHVQTHLQQPSTARGHLLVEADDVRAWSCPQHNAYRLYHCEAEQVAVCQYCCYYSGAHQGHSVCDVEIRRNEIRKMLMKQQDRLEEREQDIEDQLYKLESDKRLVEEKVNQLKEEVRLQYEKLHQLLDEDLRQTVEVLDKAQAKFCSENAAQALHLGERMQEAKKLLGSLQLLFDKTEDVSFMKNTKSVKILMDRTQTCTSSSLSPTKIGHLNSKLFLNEVAKKEKQLRKMLEGPFSTPVPFLQSVPLYPCGVSSSGAEKRKHSTAFPEASFLETSSGPVGGQYGAAGTASGEGQSGQPLGPCSSTQHLVALPGGAQPVHSSPVFPPSQYPNGSAAQQPMLPQYGGRKILVCSVDNCYCSSVANHGGHQPYPRSGHFPWTVPSQEYSHPLPPTPSVPQSLPSLAVRDWLDASQQPGHQDFYRVYGQPSTKHYVTS

Specifications

Antigen TRIM8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TRIM8 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRIM8
Gene Accession No. BC021925.1
Gene Alias GERP/RNF27
Gene Symbols TRIM8
Host Species Mouse
Immunogen TRIM8 (AAH21925.1, 1 a.a. ∼ 551 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81603
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.