Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethyl Histone H3 (Lys27) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$144.24 - $657.06
Specifications
Antigen | trimethyl Histone H3 (Lys27) |
---|---|
Applications | Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Anti-trimethyl Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated on lysine 27 (H3K73me). This purified Ab is specificity verified by dot blot (DB) and validated in western blot, immunocytochemistry, PIA. Small pack size is ideal for screening applications.Specifications
trimethyl Histone H3 (Lys27) | |
Polyclonal | |
Rabbit | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
Primary | |
2-8°C for 12 months |
Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot | |
Unconjugated | |
Human, Mouse | |
NP_003484.1 | |
Branch peptide corresponding to human Histone H3 methylated at Lys27. | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title