Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Histone H3 (Lys27), Alexa Fluor™ 647 Conjugate Rabbit anti-Human, AlexaFluor™647, Polyclonal, MilliporeSigma™

Catalog No. 07449AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-449-AF647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-449-AF647 Supplier MilliporeSigma Supplier No. 07449AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-trimethyl Histone H3 (Lys27) antibody is validated for use in immunocytochemistry for the detection of trimethyl Histone H3 (Lys27).
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl Histone H3 (Lys27)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Alexa Fluor 647
Formulation Purified rabbit conjugate antibody in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide.
Gene Symbols HIST3H3; H3FT
Host Species Rabbit
Immunogen Linear peptide corresponding to Histone H3 trimethylated at Lys27.
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.