Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 (Lys27) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07449MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-449-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-449-MI Supplier MilliporeSigma Supplier No. 07449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in western blot, Chimmunoprecipitation, ChIP-seq, immunocytochemistry, immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl Histone H3 (Lys27)
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.1M Tris-Glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide before the addition of 30% glycerol.
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...AR(me3K)SAP... in which me3K corresponds to trimethyl-lysine at residue 27 of human histone H3.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
Target Species Human, Mouse
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.