Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 (Lys36) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE435SMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE435SMI 20 μL
ABE435MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE435SMI Supplier MilliporeSigma Supplier No. ABE435S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-trimethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 36. Also known as Anti-H3K36me3, this purified Ab is specificity verified by dot blot (DB) and has been validated in western blot, immunocytochemistry, DB, Chimmunoprecipitation, ChIP-seq.
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl Histone H3 (Lys36)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 trimethylated at Lys36.
Purification Method Affinity Purified
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.