Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Histone H3 (Lys36) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE305SMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE305SMI 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE305SMI Supplier MilliporeSigma Supplier No. ABE305S

Rabbit Polyclonal Antibody

Use Anti-trimethyl Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB, immunocytochemistry, Chimmunoprecipitation to detect trimethyl Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl Histone H3 (Lys36)
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4) and 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Host Species Rabbit
Immunogen Linear peptide corresponding to human Histone H3 trimethylated at Lys36.
Purification Method Affinity Purified
Quantity 10 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003520
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.