Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 (Lys4) Rabbit anti-Human, Clone: 15-10C-E4, MilliporeSigma™

Catalog No. 05745RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-745-RMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-745-RMI Supplier MilliporeSigma Supplier No. 05745R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Use Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, Recombinant (rabbit monoclonal antibody) proven in western blot, Chimmunoprecipitation, ChIP-seq, ELISA, PIA, Mplex, DB to detect trimethyl-Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl-Histone H3 (Lys4)
Applications Multiplex, Dot Blot, ELISA, Immunoassay, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 15-10C-E4
Conjugate Unconjugated
Formulation Protein A purified, recombinant antibody in 60mM Tris, 90mM NaCl, 0.03% NaN3, 60mM Glycine, pH 7.5, 40% Glycerol.
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen BSA-conjugated, synthetic peptide containing the sequence ...RTme3KQT... in which me3K corresponds to trimethyl-lysine 4 of human histone H3.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) H3F3A, H3F3B
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.