Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethyl-Histone H4 (Lys20) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$140.00 - $668.00
Specifications
| Antigen | trimethyl Histone H4 (Lys20) |
|---|---|
| Applications | ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
07-463-SMI
|
MilliporeSigma
07463S |
10 μg |
Each for $140.00
|
|
|||||
|
07-463-MI
|
MilliporeSigma
07463 |
100 μg |
Each for $668.00
|
|
|||||
Description
Anti-trimethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of trimethyl-Histone H4 (Lys20) also known as H4K20me3, Histone H4 (tri methyl K20) and has been validated in immunoprecipitation, western blot, Chimmunoprecipitation, DB.Specifications
| trimethyl Histone H4 (Lys20) | |
| Polyclonal | |
| Rabbit | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/C; H4/D; HIST1H4J; HIST1H4D; H4/E | |
| Primary | |
| 2-8°C for 12 months |
| ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Human, Mouse, Rat | |
| NP_001029249 | |
| KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 trimethylated at Lys20. | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title