Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl(Lys9)-phospho(Ser10)-Histone H3 Rabbit anti-Human, MilliporeSigma™

Catalog No. 04809MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-809-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-809-MI Supplier MilliporeSigma Supplier No. 04809

Rabbit Monoclonal Antibody

Use Anti-trimethyl(Lys9)-phospho(Ser10)-Histone H3 Antibody (Rabbit Monoclonal Antibody) validated in western blot, PIA, DB, Mplex to detect trimethyl(Lys9)-phospho(Ser10)-Histone H3 also known as H3K9me3S10P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl(Lys9)-phospho(Ser10)-Histone H3
Applications Multiplex, Immunoassay, Inhibition Assays, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Cultured supernantant containing 0.05% sodium azide
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence ...ARme3KpST... in which me3K corresponds to trimethyl-lysine 9 and pS to phosphoserine 10 of human histone H3.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.