Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

triple functional domain (PTPRF interacting), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-352 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-352 Supplier Abnova Corporation Supplier No. H00007204A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TRIO.

Sequence: ERKSSSLKRRHYVLQELVETERDYVRDLGYVVEGYMALMKEDGVPDDMKGKDKIVFGNIHQIYDWHRDFFLGELEKCLEDPEKLGSLFVKHERRLHMYIAYCQNKPKSEH

Specifications

Antigen triple functional domain (PTPRF interacting)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TRIO.
Formulation 50% glycerol
Gene TRIO
Gene Accession No. NM_007118
Gene Alias FLJ42780/tgat
Gene Symbols TRIO
Host Species Mouse
Immunogen TRIO (NP_009049, 1961 a.a. ∼ 2070 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7204
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.