Learn More
Trk B Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB5372
Catalog No. AB5372MI
Only null left
Add to Cart
Description
Description
Anti-Trk B Antibody, pain, a.a. 54-67 rTrkB detects level of Trk B and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Trk B
Polyclonal
Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50μL of sterile distilled water. Centrifuge if necessary to remove any residue.
Rabbit
50 μL
NM_001007097.1; NM_001018064.1; NM_001018065.1; NM_001018066.1; NM_006180.3
Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Immunohistochemistry, Western Blot
Unconjugated
NTRK2; Trk-B; GP145-TrkB; TRKB; EC 2.7.10.1
A synthetic peptide from rat trkB
Primary
Human, Rat
Serum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only