Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Mouse anti-Human, Rat, Clone: 6B2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN681
Description
Anti-TrkA Antibody, clone 6B2 is a highly specific mouse monoclonal antibody, that targets TrkA and has been tested in western blotting, immunohistochemistry and immunocytochemistry.
Specifications
TrkA | |
Monoclonal | |
Unconjugated | |
NP_002520 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
6B2 | |
Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide. | |
NTRK1, MTC, TRK, TRKA | |
Purified recombinant extracellular fragment of TrkA expressed in HEK293 cells. | |
100 μL | |
Human, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction