Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Rabbit anti-Human, FITC, Polyclonal, MilliporeSigma™

Catalog No. FCABS350FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS350FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS350FMI Supplier MilliporeSigma Supplier No. FCABS350F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect TrkA using this Milli-Mark™ Anti-TrkA-FITC Antibody validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkA
Applications Flow Cytometry
Classification Polyclonal
Conjugate FITC
Formulation Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols DKFZp781I14186; EC 2.7.10.1; MTC; OTTHUMP00000038736; TRK
Host Species Rabbit
Immunogen Bacterially expressed purified protein corresponding to the entire extracellular domain of rat TrkA receptor.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) NP_067600
Target Species Human
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.