Learn More
Description
Specifications
Specifications
| Antigen | TrkA |
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit IgG in buffer containing 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide. Frozen at −20°C. |
| Gene Symbols | DKFZp781I14186; EC 2.7.10.1; MTC; OTTHUMP00000038736; TRK; TRK1; TRKA; Trk-A; p140-TrkA |
| Host Species | Rabbit |
| Immunogen | Rabbits immunized with bacterially expressed purified protein corresponding to the entire extracellular domain of rat TrkA receptor. |
| Purification Method | Protein A purfied |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.