Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkB Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9872
Description
Anti-TrkB Antibody is an antibody against TrkB for use in immunohistochemistry.
Specifications
TrkB | |
Polyclonal | |
Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative. | |
Rabbit | |
Serum | |
Primary | |
Mouse, Rat | |
Serum |
Immunohistochemistry | |
Unconjugated | |
NTRK2; Trk-B; TRKB; GP145-TrkB; EC 2.7.10.1 | |
Extracellular domain of the recombinant mouse receptor expressed in mammalian cells. | |
50 μL | |
NM_001007097.1; NM_001018064.1; NM_001018065.1; NM_001018066.1; NM_006180.3 | |
Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction