Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRNT1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89210451 Shop All Abnova Corporation Products
missing translation for 'changeView'
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-210-451 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 89-210-451 Supplier Abnova Corporation missing translation for 'supplierNo' H00051095K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human TRNT1 peptide using ARM Technology.

The CCA-adding enzyme TRNT1 (EC 2.7.7.25) is an essential enzyme that catalyzes the addition of the CCA terminus to the 3-prime end of tRNA precursors. This reaction is a fundamental prerequisite for mature tRNAs to become aminoacylated and to participate in protein biosynthesis (Lizano et al., 2007 [PubMed 17204286]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen TRNT1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description tRNA nucleotidyl transferase, CCA-adding, 1
Formulation In 1X PBS, pH 7.4
Gene TRNT1
Gene Accession No. TRNT1
Gene Alias CCA1, CGI-47, MtCCA
Gene Symbols TRNT1
Host Species Rabbit
Immunogen A synthetic peptide of human TRNT1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51095
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.