Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROP-2 Mouse anti-Human, Unlabeled, Clone: 162-46, BD

Catalog No. BDB551317 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551317 Supplier BD Biosciences Supplier No. 551317
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Trop-2/GA733-1 is a cell surface glycoprotein that was originally identified on human trophoblast and choriocarcinoma cell lines and has been shown to be expressed in most human carcinomas. Trop-2 has been cloned and shown to encode a type-1 transmembrane protein with a single transmembrane domain. Furthermore, research has shown Trop-2 to be involved in the transduction of an intracellular calcium signal, although the exact physiological mechanism has yet to be elucidated. The antibody recognizes human Trop-2. Cells from the human choriocarcinoma cell line BeWo were used as the immunogen.

Host Species: Mouse
Clone: 162-46
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human choriocarcinoma cell line BeWo

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TROP-2
Applications Flow Cytometry
Classification Monoclonal
Clone 162-46
Concentration 0.5mg/mL
Conjugate Unconjugated
Description GA733-1; TROP2; EGP-1; EGP1; GP50; M1S1; TACD2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human choriocarcinoma cell line BeWo
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.